3-(Trifluoromethyl)thiobenzamide ≥97%
Leverantör: Thermo Scientific
Fara
H26311.MD H26311.03
H26311.MDEA 635 SEK
H26311.MD H26311.03
3-(Trifluoromethyl)thiobenzamide ≥97%
3-(Trifluoromethyl)thiobenzamide
Purity: 97% CAS No.: 53515-17-6 Molecular Formula: C8H6F3NS Formula Weight: 205.20 UN No.: 2811 MDL No.: MFCD01567258
Formel: C₈H₆F₃NS
MW: 151,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01567258
CAS nummer: 53515-17-6
EINECS: None
UN: 2811
ADR: 6.1,III