β-Naphthoflavone 99+%
Leverantör: Thermo Scientific
Synonyms: 5,6-Benzoflavone, 3-Phenyl-1H-naphtho[2,1-b]pyran-1-one, 3-Phenyl-1H-benzo[f]chromen-1-one
128170010 128170050
ACRO128170010EA 930 SEK
ACRO128170010 ACRO128170050
β-Naphthoflavone 99+%
β-Naphthoflavone
Formel: C₁₉H₁₂O₂
MW: 272,3 g/mol
Kokpunkt: 461 °C (1013 hPa)
Smältpunkt: 162…164 °C
Densitet: 1,11 g/cm³ (20 °C)
MDL Number: MFCD00004986
CAS nummer: 6051-87-2
EINECS: 227-958-4
Order Now
Specifikation testresultat
Appearance Off-white to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 162 to 166 °C
HPLC ≥99.0 %