2-Mercaptobenzimidazole ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol
A18350.0E A18350.22 A18350.30 A18350.36
A18350.0EEA 5925 SEK
A18350.0E A18350.22 A18350.30 A18350.36
2-Mercaptobenzimidazole ≥97%
2-Mercaptobenzimidazole
Purity: 97% CAS No.: 583-39-1 Molecular Formula: C7H6N2S Formula Weight: 150.20 Flash Point: 250°(487°F) Melting Point: 305-310° Beilstein Reference: 119867 Merck Reference: 14,1082 UN No.: 2811 MDL No.: MFCD00466107
Formel: C₇H₆N₂S
MW: 150,2 g/mol
Kokpunkt: 376 °C (1013 hPa)
Smältpunkt: 305…310 °C
Densitet: 1,42
Flampunkt: >250 °C (487 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00466107
CAS nummer: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,1082