2-Mercaptobenzimidazole 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol
125440010 125440050 125441000 125442500
ACRO125440010EA 2100 SEK
ACRO125440010 ACRO125440050 ACRO125441000 ACRO125442500
2-Mercaptobenzimidazole 98%
2-Mercaptobenzimidazole
Formel: C₇H₆N₂S
MW: 150,2 g/mol
Kokpunkt: 376 °C (1013 hPa)
Smältpunkt: 302…307 °C
Densitet: 1,42 g/cm³ (20 °C)
Flampunkt: >250 °C
MDL Number: MFCD00466107
CAS nummer: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to beige Powder
Infrared spectrum Conforms
Ash ≤0.3 %
Titration with NaOH ≥97.5 %
Insoluble matter ≤0.3 % (in NaOH)
Volatile matter ≤0.3 %