2-Hexene cis- and trans-mixture ≥85%, Technical Grade
Leverantör: Thermo Fisher Scientific
Fara
H53472.06 H53472.14
H53472.06EA 1750 SEK
H53472.06 H53472.14
2-Hexene cis- and trans-mixture ≥85%, Technical Grade
2-Hexene
Formel: C₆H₁₂
Kokpunkt: 67…69 °C
Densitet: 0,678
Flampunkt: –20 °C (–4 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009473
CAS nummer: 592-43-8
EINECS: 209-755-2
UN: 3295
ADR: 3,II