1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
Leverantör: Merck
Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol
MQ200
8.08394.0100 8.08394.1000 8.08394.2500 8.08394.9050
8.08394.0100EA 629 SEK
8.08394.0100 8.08394.1000 8.08394.2500 8.08394.9050
1,1,1-Tris(hydroxymethyl)propane, Sigma-Aldrich®
1,1,1-Tris(hydroxymethyl)propane
Formel: C₆H₁₄O₃
MW: 134.18 g/mol
Kokpunkt: 296
Smältpunkt: 53
Densitet: 1.15 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004694
CAS nummer: 77-99-6
EINECS: 201-074-9
Order Now
Specifikation testresultat
Solubility (20 °C) soluble
Melting Point 57 - 61 °C
Molar Mass 134.17 g/mol
Bulk Density 700 kg/m3
Boiling Point 296 - 298 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (50 °C)
Flash Point 179 °C DIN 51758
Explosion Limit 2 - 11.8 %(V)
pH 6.5 (100 g/l, H2O, 20 °C) (External MSDS)
Ignition Point 375 °C DIN 51794