2-Chloroethanesulphonyl chloride 95%
Leverantör: Thermo Scientific
Fara
Synonyms: (2-Chloroethyl)sulphonyl chloride
404610250 404610050
ACRO404610250EA 1350 SEK
ACRO404610250 ACRO404610050
2-Chloroethanesulphonyl chloride 95%
2-Chloroethanesulphonyl chloride
Formel: C₂H₄Cl₂O₂S
MW: 163,02 g/mol
Kokpunkt: 78 °C (9 mmHg)
Smältpunkt: 3 °C
Densitet: 1,55 g/cm³
Flampunkt: 99 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007461
CAS nummer: 1622-32-8
EINECS: 216-594-1
UN: 2927
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
Titration Argentometric ≥94 % (ex Cl)