2,6-Difluoro-3-hydroxybenzaldehyde 95%
Leverantör: Thermo Scientific
Varning
H32250.MD H32250.03
H32250.MDEA 1900 SEK
H32250.MD H32250.03
2,6-Difluoro-3-hydroxybenzaldehyde 95%
2,6-Difluoro-3-hydroxybenzaldehyde
Purity: 95% CAS No.: 152434-88-3 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 110-114° MDL No.: MFCD08275284
Formel: C₇H₄F₂O₂
Smältpunkt: 110…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD08275284
CAS nummer: 152434-88-3