Pyridine-2,3-dicarbonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2,3-Dicyanopyridine

P1218-1G P1218-5G
TCIAP1218-1GEA 1300 SEK
TCIAP1218-1G TCIAP1218-5G
Pyridine-2,3-dicarbonitrile ≥98.0%
Pyridine-2,3-dicarbonitrile
Formel: C₇H₃N₃
MW: 129,12 g/mol
MDL Number: MFCD00191682
CAS nummer: 17132-78-4
EINECS: 249-492-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eccb2a91725727455bf9bb163d53c82d12c6bc00&vid=eccb2a91725727455bf9bb163d53c82d12c6bc00&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR