2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H,1H,5H-Octafluoro-1-pentanol
B20108.14 B20108.22
B20108.14EA 535 SEK
B20108.14 B20108.22
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Purity: 98% CAS No.: 355-80-6 Molecular Formula: F2CH(CF2)3CH2OH Formula Weight: 232.07 Flash Point: 75°(167°F) Refractive Index: 1.3180 Melting Point: -50° Boiling Point: 141-142° Beilstein Reference: 1773494 Merck Reference: 14,6746 MDL No.: MFCD00039631
Formel: C₅H₄F₈O
MW: 232,07 g/mol
Kokpunkt: 141…142 °C
Smältpunkt: < –50 °C
Densitet: 1,667
Flampunkt: 75 °C (167 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039631
CAS nummer: 355-80-6
EINECS: 206-593-4
Merck Index: 14,6746