Acetonitril, EMPLURA®, Supelco®
Leverantör: Merck
Fara
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
M-Clarity™ quality level = MQ200
1.15500.1000 1.15500.2500 1.15500.6025 1.15500.9190
1.15500.1000EA 465 SEK
1.15500.1000 1.15500.2500 1.15500.6025 1.15500.9190
Acetonitril, EMPLURA®, Supelco®
Acetonitril
Formel: H₃CCN
MW: 41.05 g/mol
Kokpunkt: 81.6 °C (1013 hPa)
Smältpunkt: –45.7 °C
Densitet: 0.782 g/cm³ (20 °C)
Flampunkt: 2 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00001878
CAS nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM