2-Bromo-4-chloro-1-fluorobenzene ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene
H25935.14 H25935.06
H25935.14EA 6250 SEK
H25935.14 H25935.06
2-Bromo-4-chloro-1-fluorobenzene ≥97%
2-Bromo-4-chloro-1-fluorobenzene
Purity: 97% CAS No.: 1996-30-1 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Flash Point: 85°(185°F) Refractive Index: 1.554 Boiling Point: 180° MDL No.: MFCD00672934
Formel: C₆H₃BrClF
MW: 209,44 g/mol
Kokpunkt: 180 °C
Densitet: 1,74
Flampunkt: 85 °C (185 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00672934
CAS nummer: 1996-30-1