2-Bromo-6-benzyloxypyridine ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 2-(Benzyloxy)-6-bromopyridine

B2576-25G B2576-5G
TCIAB2576-25GEA 6275 SEK
TCIAB2576-25G TCIAB2576-5G
2-Bromo-6-benzyloxypyridine ≥97.0%
2-Bromo-6-benzyloxypyridine
Formel: C₁₂H₁₀BrNO
MW: 264,12 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00234782
CAS nummer: 117068-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9108e9761fcc4b4c77f490bfe489a1389282eaa9&vid=9108e9761fcc4b4c77f490bfe489a1389282eaa9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR