Amantadine hydrochloride 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride
A12699.14 A12699.22 A12699.36
A12699.14EA 1225 SEK
A12699.14 A12699.22 A12699.36
Amantadine hydrochloride 99%
Amantadine hydrochloride
Purity: 99% CAS No.: 665-66-7 Molecular Formula: C10H17N.HCl Formula Weight: 187.71 Melting Point: 360° Beilstein Reference: 4198854 Merck Reference: 14,374 MDL No.: MFCD00074723
Formel: C₁₀H₁₈ClN
MW: 187,71 g/mol
Smältpunkt: 360 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074723
CAS nummer: 665-66-7
EINECS: 211-560-2
Merck Index: 14,374