3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%
Leverantör: Thermo Fisher Scientific
Varning
B22634.03 B22634.06
B22634.03EA 1100 SEK
B22634.03 B22634.06
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%
3',5'-Dimethoxy-4'-hydroxyacetophenone
Purity: 97% CAS No.: 2478-38-8 Molecular Formula: C10H12O4 Formula Weight: 196.2 Melting Point: 117-122° Beilstein Reference: 1966119
Formel: C₁₀H₁₂O₄
Kokpunkt: 335 °C (1013 hPa)
Smältpunkt: 117…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008748
CAS nummer: 2478-38-8
EINECS: 219-610-5