4'-tert-Butylacetophenone ≥98%
Leverantör: Thermo Scientific
Synonyms: p-tert-Butylacetophenone
A13615.03 A13615.06
A13615.03EA 780 SEK
A13615.03 A13615.06
4'-tert-Butylacetophenone ≥98%
4'-tert-Butylacetophenone
Purity: 98% CAS No.: 943-27-1 Molecular Formula: C12H16O Formula Weight: 176.26 Flash Point: 100°(212°F) Refractive Index: 1.5200 Melting Point: 17-18° (dimorphic) Boiling Point: 107-108°/5mm MDL No.: MFCD00017256
Formel: C₁₂H₁₆O
MW: 176,26 g/mol
Kokpunkt: 107…108 °C (5 mmHg)
Smältpunkt: 17…18 °C (dimorphic)
Densitet: 0,964
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00017256
CAS nummer: 943-27-1
EINECS: 213-399-3