Tris(perfluorobutyl)amine MS standard
Leverantör: Thermo Scientific
Varning
Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine
A19126.03 A19126.06 A19126.14
A19126.03EA 845 SEK
A19126.03 A19126.06 A19126.14
Tris(perfluorobutyl)amine MS standard
Tris(perfluorobutyl)amine
CAS No.: 311-89-7 Molecular Formula: [CF3(CF2)3]3N Formula Weight: 671.10 Refractive Index: 1.2890 Melting Point: -52° Boiling Point: 177-180° MDL No.: MFCD00000436
Formel: C₁₂F₂₇N
MW: 671,1 g/mol
Kokpunkt: 177…180 °C
Smältpunkt: –52 °C
Densitet: 1,880
Storage Temperature: Omgivande
MDL Number: MFCD00000436
CAS nummer: 311-89-7
EINECS: 206-223-1