1-(4-Methoxyphenyl)imidazole ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B25621.06 B25621.03
B25621.06EA 2625 SEK
B25621.06 B25621.03
1-(4-Methoxyphenyl)imidazole ≥98%
1-(4-Methoxyphenyl)imidazole
Purity: 98% CAS No.: 10040-95-6 Molecular Formula: C10H10N2O Formula Weight: 174.20 Odour: Odourless Melting Point: 64-67° Boiling Point: 120°/0.1mm MDL No.: MFCD00060492
Formel: C₁₀H₁₀N₂O
Kokpunkt: 120 °C (0,1 mmHg)
Smältpunkt: 64…67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00060492
CAS nummer: 10040-95-6