Trimethyl orthobenzoate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene
172920100 172920500
ACRO172920100EA 905 SEK
ACRO172920100 ACRO172920500
Trimethyl orthobenzoate 97%
Trimethyl orthobenzoate
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Kokpunkt: 87…88 °C (7 mmHg)
Flampunkt: 82 °C
MDL Number: MFCD00008474
CAS nummer: 707-07-3
EINECS: 211-897-5
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥96.0 %