(S)-(+)-Carvone (synthetic) 98%
Leverantör: Thermo Scientific
Varning
Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one
377840250 377841000
ACRO377840250EA 635 SEK
ACRO377840250 ACRO377841000
(S)-(+)-Carvone (synthetic) 98%
(S)-(+)-Carvone
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokpunkt: 230 °C (760 mmHg)
Smältpunkt: ? 15 °C
Densitet: 0,956 g/cm³
Flampunkt: 97 °C
Storage Temperature: Omgivande
MDL Number: MFCD00062997
CAS nummer: 2244-16-8
EINECS: 218-827-2
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.5010 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.955 to 0.960
Specific optical rotation +52.0° to +62.0° (20°C, 589 nm) (neat)