(R)-(-)-Carvone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: (R)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, L-p-Mentha-1(6),8-dien-2-one, (R)-(-)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one, L(-)-Carvone
A13900.0B A13900.18 A13900.30
A13900.0BEA 6800 SEK
A13900.0B A13900.18 A13900.30
(R)-(-)-Carvone ≥98%
(R)-(-)-Carvone
Purity: 98% CAS No.: 6485-40-1 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 93°(199°F) Physical Form: Liquid Density: 0.959 Refractive Index: 1.4990 Appearance: Colourless Odour: Pleasant Boiling Point: 228-230° Beilstein Reference: 2206714 Optical Rotation: -61° (neat) MDL No.: MFCD00001578
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokpunkt: 228…230 °C
Smältpunkt: 25,2 °C
Densitet: 0,959
Flampunkt: 93 °C (199 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001578
CAS nummer: 6485-40-1
EINECS: 229-352-5