(Ethylthio)acetic acid ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (Ethylthio)aceticacid
L04743.06 L04743.14
L04743.06EA 795 SEK
L04743.06 L04743.14
(Ethylthio)acetic acid ≥97%
(Ethylthio)acetic acid
Purity: 97% CAS No.: 627-04-3 Molecular Formula: CH3CH2SCH2CO2H Formula Weight: 120.18 Refractive Index: 1.4860 Boiling Point: 117-118°/11mm UN No.: 3265 MDL No.: MFCD00021769
Formel: C₄H₈O₂S
Kokpunkt: 117…118 °C (11 mmHg)
Densitet: 1,1497 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00021769
CAS nummer: 627-04-3
EINECS: 210-979-8
UN: 3265
ADR: 8,III