Bicine, Millipore®
Leverantör: Merck
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
M-Clarity™ quality level = MQ100
1.01910.0250 1.01910.9010
1.01910.0250EA 2850 SEK
1.01910.0250 1.01910.9010
Bicine, Millipore®
Bicine
Formel: C₆H₁₃NO₄
MW: 163,17 g/mol
Smältpunkt: 185…192 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004295
CAS nummer: 150-25-4
EINECS: 205-755-1