(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA
M-Clarity™ quality level = MQ200
8.40094.0100 8.40094.0010
8.40094.0100EA 1750 SEK
8.40094.0100 8.40094.0010
(1R)-(-)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(1R)-(-)-Camphor-10-sulphonic acid
Formel: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Kokpunkt: 375
Smältpunkt: 193…196 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150753
CAS nummer: 35963-20-3
EINECS: 252-817-9
UN: 3261
ADR: 8 III
Order Now
Specifikation testresultat
Solubility (20 °C) soluble
Melting Point 193 - 196 °C decomposes
Molar Mass 232.30 g/mol
pH 1.2 - 1.4 (20 g/l, H2O)