Propionaldehyd 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Propanal
131510010 131510025 131510250 131510100
ACRO131510010EA 1050 SEK
ACRO131510010 ACRO131510025 ACRO131510250 ACRO131510100
Propionaldehyd 97%
Propionaldehyd
Formel: C₃H₆O
MW: 58,08 g/mol
Kokpunkt: 48 °C (760 mmHg)
Smältpunkt: –81 °C
Densitet: 0,798 g/cm³
Flampunkt: –40 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007020
CAS nummer: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥96.0 %
UV at 260 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titration with NaOH ≥99.0 %