Caprylaldehyde för syntes, Sigma-Aldrich®

Leverantör: Merck
Varning

Synonyms: Octanal, Octylaldehyde, Octaldehyde

M-Clarity™ quality level = MQ200

8.06901.0100 8.06901.0500
8.06901.0100EA 389 SEK
8.06901.0100 8.06901.0500
Caprylaldehyde för syntes, Sigma-Aldrich®
Caprylaldehyde
Formel: C₈H₁₆O
MW: 128,21 g/mol
Kokpunkt: ∼171 °C (1013 hPa)
Smältpunkt: ∼12 °C
Densitet: 0,82 g/cm³ (20 °C)
Flampunkt: 56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007029
CAS nummer: 124-13-0
EINECS: 204-683-8
UN: 1989
ADR: 3,III
Merck Index: 13,01772

Order Now

Specifikation testresultat

Solubility 0.21 g/l (20 °C)
Melting Point 12 - 15 °C
Molar Mass 128.21 g/mol
Boiling Point 167 - 169 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 56 °C
Refractive Index 1.419 (20 °C, 589 nm)
Explosion Limit 1.0 - 6.5 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 190 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR