Caprylaldehyde 99%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Octanal, Octylaldehyde, Octaldehyde
129481000
129485000
ACRO129481000EA
525
SEK
ACRO129481000
ACRO129485000
Caprylaldehyde 99%
Caprylaldehyde
Formel:
C₈H₁₆O MW: 128,21 g/mol Kokpunkt: ∼171 °C (1013 hPa) Smältpunkt: 12…15 °C Densitet: 0,821 g/cm³ Flampunkt: 51 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007029 CAS nummer: 124-13-0 EINECS: 204-683-8 UN: 1191 ADR: 3,III Merck Index: 13,01772 |
Specifikation testresultat
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4200 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...