1-Bromo-5-chloropentane ≥98%
Leverantör: Thermo Fisher Scientific
B20479.09 B20479.18
B20479.09EA 860 SEK
B20479.09 B20479.18
1-Bromo-5-chloropentane ≥98%
1-Bromo-5-chloropentane
Purity: 98% CAS No.: 54512-75-3 Molecular Formula: Cl(CH2)5Br Formula Weight: 185.50 Flash Point: 95°(203°F) Refractive Index: 1.4836 Boiling Point: 210-212° Beilstein Reference: 1732341 MDL No.: MFCD00001016
Formel: BrH₂C(CH₂)₃CH₂Cl
Kokpunkt: 210…212 °C
Densitet: 1,408
Flampunkt: 95 °C (203 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001016
CAS nummer: 54512-75-3
EINECS: 259-193-7