Tetrabromomethane, (max. 6% H₂O) 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Carbon tetrabromide
L00821.14 L00821.22 L00821.36
L00821.14EA 870 SEK
L00821.14 L00821.22 L00821.36
Tetrabromomethane, (max. 6% H₂O) 98%
Tetrabromomethane
Purity: 98% CAS No.: 558-13-4 Molecular Formula: CBr4 Formula Weight: 331.63 Melting Point: 90-95° Boiling Point: 190° UN No.: 2516 MDL No.: MFCD00000117
Formel: Br₄C
MW: 331,63 g/mol
Kokpunkt: 190 °C
Smältpunkt: 90…95 °C
Densitet: 3,420
Storage Temperature: Omgivande
MDL Number: MFCD00000117
CAS nummer: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III