1-Bromoundecane 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Undecyl bromide
224010250 224011000
ACRO224010250EA 860 SEK
ACRO224010250 ACRO224011000
1-Bromoundecane 98%
1-Bromoundecane
Formel: BrH₂C(CH₂)₉CH₃
MW: 235,21 g/mol
Kokpunkt: 137…138 °C (18 mmHg)
Smältpunkt: –9 °C
Densitet: 1,05 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000223
CAS nummer: 693-67-4
EINECS: 211-757-3
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
GC ≥97.5 %