1-Bromohexane 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Hexyl bromide
107040025 107045000 107040100
ACRO107040025EA 2425 SEK
ACRO107040025 ACRO107045000 ACRO107040100
1-Bromohexane 99+%
1-Bromohexane
Formel: BrH₂C(CH₂)₄CH₃
MW: 165,07 g/mol
Kokpunkt: 154…158 °C (760 mmHg)
Smältpunkt: –85 °C
Densitet: 1,17 g/cm³
Flampunkt: 57 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000271
CAS nummer: 111-25-1
EINECS: 203-850-2
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4465 to 1.4485 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≤0.5 % Hexanol
Color scale ≤50 APHA
Acidity ≤0.1 % (as HBr)