1-Bromohexadecane ≥98%
Leverantör: Thermo Scientific
A15147.0B A15147.14 A15147.30
A15147.0BEA 3250 SEK
A15147.0B A15147.14 A15147.30
1-Bromohexadecane ≥98%
1-Bromohexadecane
Purity: 98% CAS No.: 112-82-3 Molecular Formula: CH3(CH2)15Br Formula Weight: 305.35 Flash Point: 176°(348°F) Refractive Index: 1.4620 Odour: Odourless Melting Point: 16-18° Boiling Point: 336° Beilstein Reference: 773989 MDL No.: MFCD00000230
Formel: BrH₂C(CH₂)₁₄CH₃
MW: 305,34 g/mol
Kokpunkt: 336 °C
Smältpunkt: 16…18 °C
Densitet: 1,000
Flampunkt: 176 °C (348 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000230
CAS nummer: 112-82-3
EINECS: 204-008-7