1-Bromohexadecane 97%

Leverantör: Thermo Scientific

215430010 215432500 215430025 215430000
ACRO215430010EA 2825 SEK
ACRO215430010 ACRO215432500 ACRO215430025 215430000.
1-Bromohexadecane 97%
1-Bromohexadecane
Formel: BrH₂C(CH₂)₁₄CH₃
MW: 305,34 g/mol
Kokpunkt: 336 °C (760 mmHg)
Smältpunkt: 17,3 °C
Densitet: 0,999 g/cm³
Flampunkt: 177 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000230
CAS nummer: 112-82-3
EINECS: 204-008-7

Order Now

Specifikation testresultat

Appearance Clear colorless to yellow to amber to brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR