1-Bromodecane 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Decyl bromide
163600010 163605000
ACRO163600010EA 2050 SEK
ACRO163600010 ACRO163605000
1-Bromodecane 98%
1-Bromodecane
Formel: BrH₂C(CH₂)₈CH₃
MW: 221,18 g/mol
Kokpunkt: 238 °C (1013 hPa)
Smältpunkt: –29,6 °C
Densitet: 1,06 g/cm³
Flampunkt: 94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000221
CAS nummer: 112-29-8
EINECS: 203-955-3
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC 97.5 %