1,2,3,4-Tetrahydro-1,1,4,4-tetramethylnaphthalene
Leverantör: Thermo Scientific
Synonyms:
1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
TL00201EA
TL00201EE
MAYBTL00201EAEA
0
SEK
MAYBTL00201EA
MAYBTL00201EE
1,2,3,4-Tetrahydro-1,1,4,4-tetramethylnaphthalene
1,2,3,4-Tetrahydro-1,1,4,4-tetramethylnaphthalene
Formel:
C₁₄H₂₀ MW: 188,31 g/mol |
MDL Number:
MFCD00052728 CAS nummer: 6683-46-1 EINECS: 229-723-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...