1,2,3,4-Tetrahydronaftalen 98+%, pure
Leverantör: Thermo Fisher Scientific
Varning
146730010 146730025 146730100
ACRO146730010EA 1850 SEK
ACRO146730010 ACRO146730025 ACRO146730100
1,2,3,4-Tetrahydronaftalen 98+%, pure
1,2,3,4-Tetrahydronaftalen
Formel: C₁₀H₁₂
MW: 132.21 g/mol
Kokpunkt: 207 °C (760 mmHg)
Smältpunkt: –35 °C
Densitet: 0.973 g/cm³
Flampunkt: 77 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001733
CAS nummer: 119-64-2
EINECS: 204-340-2
UN: 3082
ADR: 9 III
Merck Index: 12,09360
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.0 %