Lantan(III)hexahydrat ≥99.9% (REO, rare earth oxide basis)
Leverantör: Thermo Scientific
Fara
12915.36 12915.22
12915.36EA 5200 SEK
12915.36 12915.22
Lantan(III)hexahydrat ≥99.9% (REO, rare earth oxide basis)
Lantan(III)hexahydrat
Purity: 99.9% CAS No.: 10277-43-7 Molecular Formula: La(NO3)3.6H2O Formula Weight: 433.01 (324.92anhy) Physical Form: Crystalline Odour: Odourless Merck Reference: 14,5363 UN No.: 1477 MDL No.: MFCD00149751
Formel: La(NO₃)₃·6H₂O
MW: 433,01 g/mol
Kokpunkt: 126 °C (1013 hPa)
Smältpunkt: 40 °C
Densitet: 2,347 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149751
CAS nummer: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,5363