Lantan(III)hexahydrat 98+%
Leverantör: Thermo Fisher Scientific
Fara
218711000 218715000
ACRO218711000EA 1850 SEK
ACRO218711000 ACRO218715000
Lantan(III)hexahydrat 98+%
Lantan(III)hexahydrat
Formel: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Kokpunkt: 126
Smältpunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149751
CAS nummer: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1 II
Merck Index: 14,05363
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric ≥98.0 %