Lantan(III)hexahydrat 99.995%
Leverantör: Thermo Fisher Scientific
Fara
193280050 193281000 193285000
ACRO193280050EA 605 SEK
ACRO193280050 ACRO193281000 ACRO193285000
Lantan(III)hexahydrat 99.995%
Lantan(III)hexahydrat
Formel: La(NO₃)₃·6H₂O
Kokpunkt: 126 °C (1013 hPa)
Smältpunkt: 40 °C
Densitet: 2,347 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149751
CAS nummer: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,II
Merck Index: 14,05363
Order Now
Specifikation testresultat
Appearance White Crystals and/or chunks
Total trace metal impurities ≤0.005 %
Lanthanum (La) 31.0 to 35.0 %