Lantan(III)klorid heptahydrat ACS
Leverantör: Thermo Fisher Scientific
Varning
12916.18 12916.30
12916.18EA 1650 SEK
12916.18 12916.30
Lantan(III)klorid heptahydrat ACS
Lantan(III)klorid heptahydrat
CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Physical Form: Crystalline Odour: Odourless Melting Point: 91° dec. MDL No.: MFCD00149756
Formel: LaCl₃·7H₂O
MW: 371.37 g/mol
Kokpunkt: ∼860 °C (1013 hPa)
Smältpunkt: 91 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149756
CAS nummer: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363