Lantan(III)klorid heptahydrat ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
11268.18 11268.30 11268.A1
11268.18EA 2225 SEK
11268.18 11268.30 11268.A1
Lantan(III)klorid heptahydrat ≥99.99% (REO, rare earth oxide basis), REacton®
Lantan(III)klorid heptahydrat
Purity: 99.99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Physical Form: Wet Crystalline Aggregates Odour: Odourless Merck Reference: 14,5363 MDL No.: MFCD00149756
Formel: LaCl₃·7H₂O
MW: 371.37 g/mol
Kokpunkt: ∼860 °C (1013 hPa)
Smältpunkt: 91 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149756
CAS nummer: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363