Tenn(II)klorid dihydrat 98+%
Leverantör: Thermo Scientific
Fara
Synonyms: Stannous chloride dihydrate
222661000 222665000 222660025
ACRO222661000EA 605 SEK
ACRO222661000 ACRO222665000 ACRO222660025
Tenn(II)klorid dihydrat 98+%
Tin (II) chloride dihydrate
Formel: SnCl₂·2H₂O
MW: 225,65 g/mol
Kokpunkt: 652 °C (760 mmHg)
Smältpunkt: 37…38 °C
Densitet: 2,71 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149863
CAS nummer: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861
Order Now
Specifikation testresultat
Appearance Colorless to white Crystalline powder or crystals and/or chunks
Titration Iodimetric 98.0 to 103.0 %