Aluminiumkloridhexahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
|
Fara
|
Synonyms:
Aluminum chloride hexahydrate
1.01084.1000
1.01084.9026
1.01084.1000EA
3950
SEK
1.01084.1000
1.01084.9026
Aluminiumkloridhexahydrat, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®
Aluminiumklorid hexahydrat
Formel:
AlCl₃·6H₂O MW: 241.43 g/mol Smältpunkt: ∼100 °C Densitet: 2.39 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00149134 CAS nummer: 7784-13-6 EINECS: 231-208-1 UN: 3260 ADR: 8 II |