Aluminiumklorid hexahydrat 99%, ren
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Aluminum chloride hexahydrate
217470010 217475000 217470025 217470100
ACRO217470010EA 2325 SEK
ACRO217470010 ACRO217475000 ACRO217470025 ACRO217470100
Aluminiumklorid hexahydrat 99%, ren
Aluminiumklorid hexahydrat
Formel: AlCl₃·6H₂O
Smältpunkt: ~ 100 °C
Densitet: 2,39 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00149134
CAS nummer: 7784-13-6
EINECS: 231-208-1
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to yellow Adhering powder or crystals
Titration Argentometric ≥98.5 %
Water 42 to 48 % (K.F.)