Etyljodid ≥98% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Ethyl iodide
Stabilization: Stabilised with copper
A14444.0E A14444.22 A14444.36
A14444.0EEA 13925 SEK
A14444.0E A14444.22 A14444.36
Etyljodid ≥98% stabilised
Etyljodid
Stabilized with copper
Purity: 98+% CAS No.: 75-03-6 Molecular Formula: CH3CH2I Formula Weight: 155.97 Flash Point: 73°(163°F) Refractive Index: 1.5120 Melting Point: -108° Boiling Point: 70-73° Beilstein Reference: 505934 Merck Reference: 14,3813 MDL No.: MFCD00001091
Formel: C₂H₅I
MW: 155,97 g/mol
Kokpunkt: 70…73 °C
Smältpunkt: –108 °C
Densitet: 1,95
Flampunkt: >73 °C (163 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001091
CAS nummer: 75-03-6
EINECS: 200-833-1
UN: 2810
ADR: 6.1,III
Merck Index: 14,3813