Kalciumnitrat tetrahydrat ≥99.98%
Leverantör: Thermo Scientific
|
Fara
|
30482.14
30482.22
30482.14EA
1850
SEK
30482.14
30482.22
Kalciumnitrat tetrahydrat ≥99.98%
Kalciumnitrat tetrahydrat
Purity: 99.98%
CAS No.: 13477-34-4
Molecular Formula: Ca(NO3)2.4H2O
Formula Weight: 236.15 (164.09anhy)
Physical Form: Crystalline
Density: 1.82
Melting Point: 42°
Boiling Point: 132° dec.
Merck Reference: 14,1682
UN No.: 1454
MDL No.: MFCD00149604
Formel:
Ca(NO₃)₂·4H₂O MW: 236,15 g/mol Kokpunkt: 132 °C (dec.) Smältpunkt: 42 °C Densitet: 1,82 Storage Temperature: Omgivande |
MDL Number:
MFCD00149604 CAS nummer: 13477-34-4 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 14,1682 |