Strontium hexahydrat ≥99.9965%, Puratronic®
Leverantör: Thermo Scientific
Fara
10877.06 10877.14
10877.06EA 2150 SEK
10877.06 10877.14
Strontium hexahydrat ≥99.9965%, Puratronic®
Strontium hexahydrat
Purity: 99.9965% CAS No.: 10025-70-4 Molecular Formula: SrCl2.6H2O Formula Weight: 266.62 (158.53anhy) Physical Form: Powder Density: 1.93 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,8840 MDL No.: MFCD00149865
Formel: SrCl₂·6H₂O
MW: 266,62 g/mol
Kokpunkt: 100 °C (1013 hPa)
Smältpunkt: 61 °C
Densitet: 1,93
Storage Temperature: Omgivande
MDL Number: MFCD00149865
CAS nummer: 10025-70-4
EINECS: 233-971-6
Merck Index: 14,8840