Magnesiumklorid hexahydrat 99.0-102.0% ACS
Leverantör: Thermo Fisher Scientific
36226.36 36226.A3
36226.36EA 1525 SEK
36226.36 36226.A3
Magnesiumklorid hexahydrat 99.0-102.0% ACS
Magnesiumklorid hexahydrat
Purity: 99.0-102.0% CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Crystalline Density: 1.570 Melting Point: 117° dec. Merck Reference: 14,5662 MDL No.: MFCD00149781
Formel: MgCl₂·6H₂O
Kokpunkt: 1412 °C (1013 hPa)
Smältpunkt: 117 °C (dec.)
Densitet: 1,570
Storage Temperature: Omgivande
MDL Number: MFCD00149781
CAS nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662