Bariumklorid dihydrat ACS
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Barium dichloride
12310.36 12310.A3
12310.36EA 905 SEK
12310.36 12310.A3
Bariumklorid dihydrat ACS
Bariumklorid dihydrat
CAS No.: 10326-27-9 Molecular Formula: BaCl2.2H2O Formula Weight: 244.28 (208.25anhy) Physical Form: Crystalline Density: 3.097 Odour: Odourless Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00149154
Formel: BaCl₂·2H₂O
MW: 244.27 g/mol
Kokpunkt: 1560
Smältpunkt: 113 °C (-2H₂O)
Densitet: 3.097
Storage Temperature: Omgivande
MDL Number: MFCD00149154
CAS nummer: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1 III
Merck Index: 14,971