Natriumselenat Dekahydrat ≥99.9%
Leverantör: Thermo Scientific
Fara
14239.14 14239.22
14239.14EA 2425 SEK
14239.14 14239.22
Natriumselenat Dekahydrat ≥99.9%
Natriumselenat Dekahydrat
Purity: 99.9% CAS No.: 10102-23-5 Molecular Formula: Na2SeO4.10H2O Formula Weight: 369.09 (188.94anhy) Physical Form: Crystalline Density: 1.600 Melting Point: 35° dec. Solubility: Soluble in water Merck Reference: 14,8671 UN No.: 2630 MDL No.: MFCD00149174
Formel: Na₂SeO₄·10H₂O
MW: 369,09 g/mol
Smältpunkt: 35 °C (dec.)
Densitet: 1,600
Storage Temperature: Omgivande
MDL Number: MFCD00149174
CAS nummer: 10102-23-5
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Merck Index: 14,8671